Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-10 Rat anti-Human, BV421, Clone: JES3-9D7, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 566276
Description
IL-10 Monoclonal antibody specifically detects IL-10 in Human samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications
IL-10 | |
Monoclonal | |
Brilliant Violet 421 | |
CSIF; Cytokine synthesis inhibitory factor; TGIF | |
Recombinant Human IL-10 | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. | |
IgG1 |
Flow Cytometry, Flow Cytometry (Intracellular Staining) | |
JES3-9D7 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
25 Tests | |
Cell Biology | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction