Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-10 Rat anti-Human, PE, Clone: JES3-19F1, BD

Catalog No. BDB562035 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562035 25 Tests
BDB559330 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB562035 Supplier BD Biosciences Supplier No. 562035
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The JES3-19F1 antibody reacts with human interleukin-10 (IL-10). The immunogen used to generate the JES3-19F1 hybridoma was recombinant human IL-10 expressed in COS cells. This is a neutralizing antibody. This antibody also reacts with viral IL-10.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-10
Applications Flow Cytometry
Classification Monoclonal
Clone JES3-19F1
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Recombinant Human IL-10
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Virus
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.