Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-10 Rat anti-Human, Virus, APC-R700, Clone: JES3-19F1, BD Biosciences™

Catalog No. BDB567012 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB567012 100 Tests
BDB567013 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB567012 Supplier BD Biosciences Supplier No. 567012
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-10 Monoclonal antibody specifically detects IL-10 in Human, Virus samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-10
Applications Flow Cytometry
Classification Monoclonal
Clone JES3-19F1
Conjugate APC-R700
Formulation Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide.
Gene Alias Interleukin-10; CSIF; Cytokine synthesis inhibitory factor; TGIF
Host Species Rat
Immunogen Recombinant Human IL-10
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Virus
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.