Learn More
IL-10 Rat anti-Mouse, PE, Clone: JES5-16E3, BD
Rat Monoclonal Antibody
$116.50 - $262.00
Specifications
| Antigen | IL-10 |
|---|---|
| Clone | JES5-16E3 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
The JES5-16E3 monoclonal antibody specifically binds to the mouse cytokine, Interleukin-10 (IL-10). IL-10 is also known as Cytokine Synthesis Inhibitory Factor (CSIF). It is produced by various activated cell types including CD4+ T cells, CD8+ T cells, T regulatory cells, NK T cells, B1 B cells, NK cells, macrophages, dendritic cells, mast cells, granulocytes and keratinocytes. IL-10 plays a pivotal role in regulating immune responses and protecting the host from damage caused by inflammatory and autoimmune responses. IL-10 has numerous biological activities including the inhibition of cytokine synthesis by activated T cells, NK cells, monocytes, and macrophages. In the presence of accessory cells, IL-10 inhibits mitogen- or anti-CD3 induced proliferation of T lymphocytes. IL-10 has also been shown to costimulate the development of thymocytes, B cell differentiation and the generation of cytotoxic T cells. The immunogen used to generate the JES5-16E3 hybridoma was recombinant mouse IL-10. JES5-16E3 is a neutralizing antibody.
Specifications
| IL-10 | |
| 0.2mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Recombinant mouse IL-10 | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| JES5-16E3 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG2b | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.