Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-10R alpha Antibody (1B1.3a) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP308221
Description
IL-10R alpha Monoclonal specifically detects IL-10R alpha in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
IL-10R alpha | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha | |
Purified recombinant extracellular region of Mouse IL-10R alpha | |
100 μg | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Immunofluorescence | |
1B1.3a | |
Unconjugated | |
10mM PBS | |
Rat | |
Protein A or G purified | |
RUO | |
3587 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction