Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-10R alpha Antibody (1B1.3a) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | IL-10R alpha |
---|---|
Clone | 1B1.3a |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry 1:10-1:100 |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017552
![]() |
Novus Biologicals
NB100635770.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10063577
![]() |
Novus Biologicals
NB10063577 |
0.125 mg |
Each for $499.50
|
|
|||||
Description
IL-10R alpha Monoclonal specifically detects IL-10R alpha in Mouse, Human (Negative) samples. It is validated for Flow Cytometry.Specifications
IL-10R alpha | |
1.0 mg/mL | |
Flow Cytometry | |
Unconjugated | |
Rat | |
Human (Negative), Mouse | |
CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha | |
IL10RA | |
IgG1 | |
Protein G purified | |
NB100-63577 recognizes mouse CDw210a, a cell surface glycoprotein of approximately 110KD, also known as Interleukin-10 receptor 1 (IL-10R1). The epitope recognised by this antibody has been localised to the ligand binding domain within the extracellular region of mouse CDw210a. Mouse CDw210a is expressed by a range of cells including thymocytes, T cells, B cells and monocytes. This clone is reported to neutralize the effects of IL-10 (1). |
1B1.3a | |
Flow Cytometry 1:10-1:100 | |
Monoclonal | |
Purified | |
RUO | |
Q61727 | |
3587 | |
Purified recombinant extracellular region of mouse CD210 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title