Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-10R alpha Antibody (CL8203), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP32116425UL
Description
IL-10R alpha Monoclonal antibody specifically detects IL-10R alpha in Human samples. It is validated for Western BlotSpecifications
| IL-10R alpha | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| CL8203 | |
| Western Blot 1 ug/ml | |
| CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha | |
| This antibody was developed against a Synthetic Peptide of Q13651. Actual immunogen sequence is proprietary. | |
| 25 μg | |
| Cytokine Research | |
| 3587 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction