Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-10R alpha Antibody (RM0101-9E12) - Azide and BSA Free, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | IL-10R alpha |
---|---|
Clone | RM0101-9E12 |
Concentration | LYOPH |
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Description
IL-10R alpha Monoclonal specifically detects IL-10R alpha in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IL-10R alpha | |
LYOPH | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha | |
IL10RA | |
Primary | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a hamster immunized with mouse recombinant protein of extracellular domain of IL-10R alpha. |
RM0101-9E12 | |
Western Blot 1:500-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
Mouse | |
3587 | |
Mouse recombinant protein of the Extracellular domain of IL-10R alpha | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title