Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-10R alpha Antibody (RM0101-9E12), DyLight 405, Novus Biologicals™

Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP122459V
Description
IL-10R alpha Monoclonal antibody specifically detects IL-10R alpha in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
IL-10R alpha | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IL10RA | |
Mouse recombinant protein of the Extracellular domain of IL-10R alpha | |
0.1 mL | |
Primary | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a hamster immunized with mouse recombinant protein of extracellular domain of IL-10R alpha. | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0101-9E12 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha | |
Hamster | |
Protein A or G purified | |
Cytokine Research | |
3587 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction