Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-10RB Polyclonal Antibody, Huabio

Catalog No. 502544549
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4549 Supplier HUABIO RESEARCH INC Supplier No. HA500322
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IL-10RB Polyclonal antibody specifically detects IL-10RB in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen IL-10RB
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene IL10RB
Gene Accession No. Q08334
Gene Alias 6620401D04Rik; AI528744; CDW210B; CRF2-4; CRFB4; cytokine receptor class-II CRF2-4; cytokine receptor class-II member 4; Cytokine receptor family 2 member 4; cytokine receptor family II, member 4; D16H21S58; D21S58; D21S58h; D21S66; IL-10 receptor subunit beta; IL-10R subunit 2; IL-10R subunit beta; Il10r2; IL-10R2; IL10RB; IL-10RB; inte; interleukin 10 receptor 2; interleukin 10 receptor subunit beta; interleukin 10 receptor, beta; interleukin-10 receptor subunit 2; Interleukin-10 receptor subunit beta; RGD1560373
Host Species Rabbit
Immunogen Synthetic peptide within human IL-10RB aa 170-220 (Extracellular domain).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3588
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.