Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-11 Antibody (MM0361-9T27), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211655F
Description
IL-11 Monoclonal antibody specifically detects IL-11 in Human samples. It is validated for Western Blot, NeutralizationSpecifications
IL-11 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Neutralization | |
MM0361-9T27 | |
Western Blot, Block/Neutralize | |
Adipogenesis inhibitory factor, AGIFoprelvekin, IL-11Oprelvekin, interleukin 11, interleukin-11 | |
Human recombinant IL-11 | |
0.1 mL | |
Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity | |
3589 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction