Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-11 Antibody (RM0256-22C39), HRP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP211656H
Description
IL-11 Monoclonal specifically detects IL-11 in Mouse samples. It is validated for Western Blot.Specifications
IL-11 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
IL11 | |
Mouse recombinant IL-11 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot | |
RM0256-22C39 | |
Western Blot | |
Adipogenesis inhibitory factor, AGIFoprelvekin, IL-11Oprelvekin, interleukin 11, interleukin-11 | |
Rat | |
Protein A or G purified | |
Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity | |
3589 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction