Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-11 Antibody (MM0361-9T27) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | IL-11 |
---|---|
Clone | MM0361-9T27 |
Concentration | LYOPH |
Applications | Western Blot, Blocking Assay |
Classification | Monoclonal |
Description
IL-11 Monoclonal specifically detects IL-11 in Human samples. It is validated for Western Blot, Block/Neutralize.Specifications
IL-11 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
P20809 | |
3589 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
MM0361-9T27 | |
Western Blot, Blocking Assay | |
Unconjugated | |
Mouse | |
Human | |
Adipogenesis inhibitory factor, AGIFoprelvekin, IL-11Oprelvekin, interleukin 11, interleukin-11 | |
IL11 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title