Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-11R alpha Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298219B
Description
IL-11R alpha Polyclonal specifically detects IL-11R alpha in Mouse samples. It is validated for ELISA.Specifications
IL-11R alpha | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
Biotin | |
AI314697, GP130, IL-11RA, Il11ra2, NR1 | |
Produced in rabbits immunized with purified, recombinant IL-11R alpha (NP_001156873.1; Met1-Gln367). | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity | |
3590 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction