Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-12 alpha Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS0767R

Description
Interleukin-12 (IL-12) is a heterodimeric 70 kDa cytokine composed of two covalently linked, glycosylated chains with molecular weights of 40kD (p40) and 35-kD (p35). IL-12 is mainly produced by monocytes, macrophages, and dendritic cells in response to bacterial products such as lipopolysaccharides (LPS), to intracellular pathogens or upon interaction with activated T cells. IL-12 was originally discovered because of its ability to induce interferon-gamma (IFN-g) production, cell proliferation, and cytotoxicity mediated by natural killer cells and T cells. Studies have established that IL-12 also plays a key role in the development of Th1 responses, leading to IFN-g and IL-2 production. These cytokines can in turn promote T-cell responses and macrophage activation. Recombinant mouse IL-12 p70 is produced in baculovirus-infected insect cells as an authentic heterodimer of precursor p35 and p40 subunits using a dual promoter expression system. IL-12 p70 is distinct from other available forms of the protein in that it is expressed as a true heterodimer, as opposed to a single-chain, pseudo-heterodimer in which the subunits are joined by an artificial linker. The responses of lymphocytes to this cytokine are mediated by the activator of transcription protein STAT4. Nitric oxide synthase 2A (NOS2A/NOS2) is found to be required for the signaling process of this cytokine in innate immunity. Human and mouse IL-12 p40 share about 70% amino acid sequence homology.Specifications
IL-12 alpha | |
Polyclonal | |
Unconjugated | |
Il12b | |
20C2; BOS_1122; BOS_8400; CLMF; CLMF p35; CLMF p40; CLMF2; cytokine; cytotoxic lymphocyte maturation factor 1, p35; Cytotoxic lymphocyte maturation factor 35 kDa subunit; Cytotoxic lymphocyte maturation factor 40 kDa subunit; IL 12p70; Il12; IL-12; Il-12 p35; IL-12 p75; IL-12 subunit p35; IL-12 subunit p40; IL-12, subunit p35; IL12, subunit p40; Il-12; cytokine; IL12A; IL-12A; IL12A/B; IL12AB; IL12B; IL-12B; IL12P35; IL-12p35; Il12p40; Il-12p40; IL35 subunit; ILN; IMD28; IMD29; Interleukin; interleukin 12 35kDa subunit; interleukin 12 40kDa subunit; interleukin 12 p35 subunit; interleukin 12 p40 subunit precursor; interleukin 12 polypeptide 35; interleukin 12, p35; interleukin 12, p40; interleukin 12a; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin 12B; interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleukin 12p35 subunit precursor; interleukin-12 alpha chain; interleukin-12 alpha p40 subunit; interleukin-12 beta chain; interleukin-12 beta p35 subunit; interleukin-12 p35 subunit precursor; interleukin-12 p40; interleukin-12 p40 subunit; interleukin-12 p40 subunit precursor; Interleukin-12 p70; Interleukin-12 subunit alpha; Interleukin-12 subunit beta; Ll12a; MGC151228; MGC151232; natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35; natural killer cell stimulatory factor, 40 kD subunit; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NK cell stimulatory factor chain 2; NKSF; NKSF1; NKSF2; p35; p40 | |
Rabbit | |
Protein A | |
RUO | |
3592, 3593 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P29459, P29460 | |
IL12A, Il12b | |
KLH conjugated synthetic peptide derived from mouse IL-12 alpha. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction