Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-12 Antibody (1-1A4), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250440G
Description
IL-12 Monoclonal specifically detects IL-12 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
IL-12 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IL12A | |
Recombinant human IL-12 produced in Baculovirus. | |
0.1 mL | |
Immunology, Innate Immunity | |
3592 | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
1-1A4 | |
Western Blot, ELISA, Immunohistochemistry | |
CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Binds p40 subunit of IL-12 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction