Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ IL-12 Antibody Pair [HRP]
SDP

Catalog No. NB397955 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397955 5 Plates
NB397956 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB397955 Supplier Novus Biologicals™ Supplier No. NBP3116545PLATES
Add to Cart
Add to Cart

Antibody Pair

Specifications

Assay Range 31.25 to 2000 pg/mL (example only, lot dependent)
Assay Sensitivity 39.06 pg/mL (example only; lot dependent)
Conjugate HRP
Detection Method Sandwich ELISA
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Sandwich ELISA
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IL-12.
Target Species Human
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 3592
Gene Symbol IL12A
Quantity 5 Plates
Research Discipline Alzheimers Research, Immunology, Innate Immunity, Neurodegeneration
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12 A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.