Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12/IL-23 p40 Monoclonal Antibody (eBioHP40 (HP40, HP-40)), eFluor™ 660, eBioscience™, Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

$426.00

Specifications

Antigen IL-12/IL-23 p40
Concentration 5 μL/Test
Applications Flow Cytometry
Classification Monoclonal
Conjugate eFluor 660
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-164-70
GREENER_CHOICE
Encompass_Preferred
View Documents
Invitrogen™
50723542
100 Tests
Each of 1 for $426.00
Only null left
Add to Cart
 
Description

Description

Description: The eBioHP40 antibody reacts with the p40 subunit of human IL-12 p70, and human IL-23. IL-12 is a 70 kDa heterodimeric cytokine composed of two covalently linked chains, p40 and p35, and is secreted primarily by activated monocytes, macrophages, and dendritic cells. IL-23 is a biologically-related, heterodimeric cytokine composed of disulfide-linked p40 and p19 subunits. The eBioHP40 antibody has slightly lower background staining than C8.6 and recognizes a different epitope. Applications Reported: This eBioHP40 (HP40, HP-40) antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This eBioHP40 (HP40, HP-40) antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of cultured human dendritic cells. This can be used at 5 μL (0.03 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

IL-12 is an interleukin produced by dendritic cells, neutrophils, macrophages and human lymphoblastoid cells as a result of antigenic stimulation. IL-12 is a heterodimeric cytokine encoded by IL-12A (p35) and IL-12B (p40). IL-23 is a heterodimeric cytokine that shares a subunit with IL-12 (IL-12B) along with an IL-23A (IL-23p19) subunit. Functionally, IL-12 increases the expression of vascular endothelial adhesion molecule-1 on the endothelium, directs leukocytes towards tumors and facilitates the ischemic-hemorrhagic necrosis of the tumor tissue. In comparison, IL-23 plays an important role in stimulating memory T-cells, and is required for the induction, expansion, maintenance and downstream effector functions of Th17 cells, which play a vital role in upregulating neutrophil chemokines and various pro-inflammatory cytokines.
Specifications

Specifications

IL-12/IL-23 p40
Flow Cytometry
eFluor 660
Mouse
Human
Primary
4°C, store in dark, DO NOT FREEZE!
IL12A
5 μL/Test
Monoclonal
Liquid
RUO
IgG1 κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.