Learn More
Invitrogen™ IL-12/IL-23 p40 Monoclonal Antibody (31052)

Description
In ELISAs, this antibody shows less than 5% cross-reactivity with recombinant human (rh) IL-12 (p35/p40 heterodimer) and rhIL-23 (p19/p40 heterodimer). No significant cross-reactivity or interference was observed with rhIL-1 alpha, rmIL-1 alpha, rhIL-1 beta, rmIL-1 beta, rhIL-1ra, rhIL-1 RI, rhIL-1 RII, rhIL-2, rhIL-2 R alpha, rhIL-3, rmIL-3, rhIL-4, rmIL-4, rhIL-4 R, rhIL-5, rmIL-5, rhIL-5 R beta, rhIL-6, rhIL-6 R, rmIL-6, rhIL-7, rmIL-7, rhIL-8, rhIL-9, rmIL-9, rhIL-10, rmIL-10, rhIL-11, rmIL-12, rmIL-12 p40, rhIL-13, or rmIL-13. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | IL-12/IL-23 p40 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 31052 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | IL12A |
| Gene Accession No. | P29459, P29460, Q9NPF7 |
| Gene Alias | CLMF; CLMF p35; CLMF p40; CLMF2; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; Cytotoxic lymphocyte maturation factor 40 kDa subunit; il 12; IL 12/IL 23p40; il 23; IL 23 p40; Il12; Il-12 p35; IL12 p40; IL23; IL-12 subunit p35; IL-12 subunit p40; IL-12, subunit p35; IL12, subunit p40; Il12a; IL-12A; Il12b; IL-12B; IL12p35; IL-12p35; Il12p40; Il-12p40; il23; IL-23; IL23 p41; IL-23 subunit alpha; Il23a; IL-23A; IL-23-A; IL23P19; IL-23p19; IL35 subunit; ILN; IMD28; IMD29; Interleukin; Interleukin 12; interleukin 12 35 kDa subunit; interleukin 12 35kDa subunit; interleukin 12 40 kDa subunit; interleukin 12 40kDa subunit; interleukin 12 p35 subunit; interleukin 12 polypeptide 35; interleukin 12 subunit p40; interleukin 12, p35; interleukin 12, p40; interleukin 12A; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin 12b; interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleukin 23 p19 subunit; interleukin 23 subunit alpha; interleukin 23, alpha subunit p19; Interleukin12; interleukin-12 alpha chain; interleukin-12 beta chain; interleukin-12 p40; interleukin-12 p40 subunit; interleukin-12 subunit alpha; Interleukin-12 subunit beta; interleukin-12 subunit p40; interleukin-23 p19 subunit; interleukin-23 p40 subunit; interleukin-23 subunit alpha; Interleukin-23 subunit p19; interleukin-six, G-CSF related factor; JKA3 induced upon T-cell activation; Ll12a; natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35; natural killer cell stimulatory factor, 40 kD subunit; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NK cell stimulatory factor chain 2; NKSF; NKSF1; NKSF2; p19; P35; p40; RP23-388G23.1; SGRF; UNQ2498/PRO5798 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.