Learn More
Invitrogen™ IL-12/IL-23 p40 Monoclonal Antibody (C8.6), eFluor™ 660, eBioscience™

Mouse Monoclonal Antibody
$425.00
Specifications
Antigen | IL-12/IL-23 p40 |
---|---|
Clone | C8.6 |
Concentration | 5 μL/Test |
Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-712-942
![]() ![]() |
Invitrogen™
50712942 |
100 Tests |
Each of 1 for $425.00
|
|
|||||
Description
Description: The C8.6 antibody reacts with the p40 subunit of human IL-12 p70 and human IL-23. IL-12 is a 70 kDa heterodimeric cytokine composed of two covalently linked chains, p40 and p35, and secreted primarily by activated monocytes, macrophages, and dendritic cells. IL-23 is a biologically-related, heterodimeric cytokine composed of disulfide-linked p40 and p19 subunits. Applications Reported: This C8.6 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This C8.6 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of stimulated human cells. This can be used at 5 μL (0.03 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
IL-12 is an interleukin produced by dendritic cells, neutrophils, macrophages and human lymphoblastoid cells as a result of antigenic stimulation. IL-12 is a heterodimeric cytokine encoded by IL-12A (p35) and IL-12B (p40). IL-23 is a heterodimeric cytokine that shares a subunit with IL-12 (IL-12B) along with an IL-23A (IL-23p19) subunit. Functionally, IL-12 increases the expression of vascular endothelial adhesion molecule-1 on the endothelium, directs leukocytes towards tumors and facilitates the ischemic-hemorrhagic necrosis of the tumor tissue. In comparison, IL-23 plays an important role in stimulating memory T-cells, and is required for the induction, expansion, maintenance and downstream effector functions of Th17 cells, which play a vital role in upregulating neutrophil chemokines and various pro-inflammatory cytokines.Specifications
IL-12/IL-23 p40 | |
5 μL/Test | |
Flow Cytometry | |
eFluor 660 | |
Mouse | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P29459, P29460, Q9NPF7 | |
3592, 3593, 51561 | |
Primary | |
Affinity chromatography |
C8.6 | |
4°C, store in dark, DO NOT FREEZE! | |
Monoclonal | |
Liquid | |
IgG1 κ | |
Human | |
IL12A | |
CLMF; CLMF p35; CLMF p40; CLMF2; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; Cytotoxic lymphocyte maturation factor 40 kDa subunit; il 12; IL 12/IL 23p40; il 23; IL 23 p40; Il12; Il-12 p35; IL12 p40; IL23; IL-12 subunit p35; IL-12 subunit p40; IL-12, subunit p35; IL12, subunit p40; Il12a; IL-12A; Il12b; IL-12B; IL12p35; IL-12p35; Il12p40; Il-12p40; il23; IL-23; IL23 p41; IL-23 subunit alpha; Il23a; IL-23A; IL-23-A; IL23P19; IL-23p19; IL35 subunit; ILN; IMD28; IMD29; Interleukin; Interleukin 12; interleukin 12 35 kDa subunit; interleukin 12 35kDa subunit; interleukin 12 40 kDa subunit; interleukin 12 40kDa subunit; interleukin 12 p35 subunit; interleukin 12 polypeptide 35; interleukin 12 subunit p40; interleukin 12, p35; interleukin 12, p40; interleukin 12A; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin 12b; interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleukin 23 p19 subunit; interleukin 23 subunit alpha; interleukin 23, alpha subunit p19; Interleukin12; interleukin-12 alpha chain; interleukin-12 beta chain; interleukin-12 p40; interleukin-12 p40 subunit; interleukin-12 subunit alpha; Interleukin-12 subunit beta; interleukin-12 subunit p40; interleukin-23 p19 subunit; interleukin-23 p40 subunit; interleukin-23 subunit alpha; Interleukin-23 subunit p19; interleukin-six, G-CSF related factor; JKA3 induced upon T-cell activation; Ll12a; natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35; natural killer cell stimulatory factor, 40 kD subunit; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NK cell stimulatory factor chain 2; NKSF; NKSF1; NKSF2; p19; P35; p40; RP23-388G23.1; SGRF; UNQ2498/PRO5798 | |
IL12A, IL12B, Il23a | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.