Learn More
IL-12/IL-23 p40 Monoclonal Antibody (C8.6), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$95.65 - $394.00
Specifications
Antigen | IL-12/IL-23 p40 |
---|---|
Clone | C8.6 |
Concentration | 1 mg/mL |
Applications | ELISA, Functional Assay, Neutralization |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
16-712-985
![]() |
Invitrogen™
16712985 |
500 μg |
Each for $394.00
|
![]() |
|||||
50-255-0785
![]() |
Invitrogen™
16712981 |
50 μg |
Each
for $95.65
Save 25% Reg : |
![]() |
|||||
Description
Description: The C8.6 antibody reacts with the p40 subunit of human IL-12 p70 and human IL-23. IL-12 is a 70 kDa heterodimeric cytokine composed of two covalently linked chains, p40 and p35, and secreted primarily by activated monocytes, macrophages, and dendritic cells. IL-23 is a biologically-related, heterodimeric cytokine composed of disulfide-linked p40 and p19 subunits. Applications Reported: The C8.6 antibody has been reported for use ELISA, ELISPOTand neutralization. Applications Tested: The Functional Grade Purified C8.6 antibody has been tested by LAL assay to verify low endotoxin levels and has been tested for ELISA detection and in bioassay for neutralization of IL-12 p70 and IL-23 bioactivity. The C8.6 antibody at 60.0 ng/mL has been found to inhibit by 50% the biological effects of 1 ng/mL human IL-12 p70 (14-8129) or human IL-23 (14-8239), in corresponding bioassay. Detailed information and protocols about cytokine bioassays and in vitro cytokine neutralization using antibodies can be found in the BestProtocols® section. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
IL-12 is an interleukin produced by dendritic cells, neutrophils, macrophages and human lymphoblastoid cells as a result of antigenic stimulation. IL-12 is a heterodimeric cytokine encoded by IL-12A (p35) and IL-12B (p40). IL-23 is a heterodimeric cytokine that shares a subunit with IL-12 (IL-12B) along with an IL-23A (IL-23p19) subunit. Functionally, IL-12 increases the expression of vascular endothelial adhesion molecule-1 on the endothelium, directs leukocytes towards tumors and facilitates the ischemic-hemorrhagic necrosis of the tumor tissue. In comparison, IL-23 plays an important role in stimulating memory T-cells, and is required for the induction, expansion, maintenance and downstream effector functions of Th17 cells, which play a vital role in upregulating neutrophil chemokines and various pro-inflammatory cytokines.Specifications
IL-12/IL-23 p40 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
IgG1 κ | |
Affinity chromatography |
C8.6 | |
ELISA, Functional Assay, Neutralization | |
Functional Grade | |
Mouse | |
Human | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.