Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-12/IL-23 p40 Antibody (54), DyLight 405

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289244V
Description
IL-12/IL-23 p40 Monoclonal specifically detects IL-12/IL-23 p40 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
IL-12/IL-23 p40 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
3593 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
54 | |
ELISA, Sandwich ELISA Detection | |
CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2IL12, subunit p40 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-12/IL-23 p40 (Met 1-Ser 328; NP_002178.2) . | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction