Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12/IL-23 p40 Monoclonal Antibody (C17.8), Alexa Fluor™ 488, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Supplier:  Invitrogen 53712382

Encompass_Preferred

Catalog No. 50-112-4613


Add to Cart

Description

Description

Description: The C17.8 antibody reacts with the p40 subunit of IL-12 and IL-23. The C17.8 antibody has been found to neutralize IL-12 and IL-23 bioactivity. For specific neutralization of IL-23 only, please see clone G23-8 (anti-mouse IL-23 p19). IL-12 is a 70 kDa heterodimeric cytokine composed of two covalently linked chains, p40 and p35, and is secreted by activated monocytes, macrophages, and dendritic cells. IL-23 is a heterodimer composed of the disulfide-linked p19 protein and p40 protein, expressed by activated dendritic cells. Applications Reported: This C17.8 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This C17.8 antibody has been tested by intracellular staining and flow cytometric analysis of stimulated mouse cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

IL-12/IL-23 p40 Monoclonal antibody specifically detects IL-12/IL-23 p40 in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-12/IL-23 p40
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P43431, P43432, Q9EQ14
IL12A, IL12B, Il23a
Affinity chromatography
RUO
16159, 16160, 83430
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
C17.8
Alexa Fluor 488
Il23a
CLMF; CLMF p35; CLMF p40; CLMF2; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; Cytotoxic lymphocyte maturation factor 40 kDa subunit; il 12; IL 12/IL 23p40; il 23; IL 23 p40; Il12; Il-12 p35; IL12 p40; IL23; IL-12 subunit p35; IL-12 subunit p40; IL-12, subunit p35; IL12, subunit p40; Il12a; IL-12A; Il12b; IL-12B; IL12p35; IL-12p35; Il12p40; Il-12p40; il23; IL-23; IL23 p41; IL-23 subunit alpha; Il23a; IL-23A; IL-23-A; IL23P19; IL-23p19; IL35 subunit; ILN; IMD28; IMD29; Interleukin; Interleukin 12; interleukin 12 35 kDa subunit; interleukin 12 35kDa subunit; interleukin 12 40 kDa subunit; interleukin 12 40kDa subunit; interleukin 12 p35 subunit; interleukin 12 polypeptide 35; interleukin 12 subunit p40; interleukin 12, p35; interleukin 12, p40; interleukin 12A; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin 12b; interleukin 12B (natural killer cell stimulatory f
Rat
100 μg
Primary
Mouse
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.