Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-12/IL-35 p35 Antibody (116219), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1171M
Description
IL-12/IL-35 p35 Monoclonal antibody specifically detects IL-12/IL-35 p35 in Porcine samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
IL-12/IL-35 p35 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry, CyTOF | |
116219 | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
CLMF,IL12A,IL-12A,IL-12p35,Ll12a,NFSK,NKSF1,P35 | |
Chinese hamster ovary cell line CHO-derived recombinant porcine IL-12 p35/p40 heterodimer, p40 Ile23-Asn324, p35 Arg26-Ser222, Accession # Q28938, Q29053 | |
0.5 mL | |
Innate Immunity | |
Pig | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction