Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    IL-12/IL-35 p35 Antibody (2038C), mFluor Violet 500 SE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB15701MFV500
Description
IL-12/IL-35 p35 Monoclonal antibody specifically detects IL-12/IL-35 p35 in Human samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
| IL-12/IL-35 p35 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Innate Immunity | |
| Human | |
| IgG | 
| Flow Cytometry, Immunocytochemistry | |
| 2038C | |
| Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
| CLMF;IL12A;IL-12A;IL-12p35;Ll12a;NFSK;NKSF1;P35 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-12 p40 and human IL-12 p35, Ile23-Ser328 of p40, Arg23-Ser219 of p35, Accession # P29460 (p40) and P29459 (P35) | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            