Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-12/IL-35 p35 Antibody (B-T21), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314604JF646
Description
IL-12/IL-35 p35 Monoclonal antibody specifically detects IL-12/IL-35 p35 in Human samples. It is validated for ELISASpecifications
IL-12/IL-35 p35 | |
Monoclonal | |
Janelia Fluor 646 | |
CLMF;IL12A;IL-12A;IL-12p35;Ll12a;NFSK;NKSF1;P35 | |
Recombinant human IL-12/IL-35 p35 | |
0.1 mL | |
Innate Immunity | |
Human | |
Purified |
ELISA | |
B-T21 | |
50mM Sodium Borate | |
Mouse | |
Affinity purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction