Learn More
Invitrogen™ IL-12 p35 Monoclonal Antibody (B-T21 (BT21)), eBioscience™

Mouse Monoclonal Antibody
$231.65
Specifications
Antigen | IL-12 p35 |
---|---|
Clone | B-T21 (BT21) |
Concentration | 0.5 mg/mL |
Content And Storage | 4°C |
Applications | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-112-8691
![]() ![]() |
Invitrogen™
14712882 |
100 μg |
Each of 1
for $231.65
Save 12% Reg : |
|
|||||
Description
Description: The B-T21 antibody reacts with the p35 subunit of human interleukin-12 (IL-12). IL-12 is a 70 kDa heterodimeric cytokine composed of two covalently linked chains, p40 and p35, and is secreted primarily by activated B cells. For optimal detection of human IL-12 p70 by ELISA, a combination of purified B-T21 antibody for capture and biotinylated C8.6 antibody for detection is recommended. Applications Reported: The B-T21 antibody has been reported for use as capture antibody in an IL-12 p70 sandwich ELISA. Applications Tested: The B-T21 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of human IL-12 p70 in combination with the biotinylated C8.6 (13-7129) antibody for detection and recombinant human IL-12 p70 (14-8129) as the standard. A suitable range of concentrations of this antibody for ELISA capture is 1 - 4 μg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 2000 pg/mL - 15 pg/mL should be included in each ELISA plate. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Interleukin 12A is a novel signaling cytokine belonging to the Interleukin-6 protein superfamily. Interleukin-12 alpha chain precursor (IL-12A) is a 35-kD subunit of disulfide-linked heterodimer of cytokine IL-12 and has a broad array of biological activities. Reports suggest that IL-12A can act as a growth factor for activated T and NK cells and enhance the lytic activity of NK/lymphokine-activated Killer cells. IL-12A stimulates the production of IFN-gamma by resting PBMC as well as T-cell-independent induction of interferon (IFN)-gamma, and helps in the differentiation of both Th1 and Th2 cells. It is also involved in positive regulation of tyrosine phosphorylation of Stat4 protein. The signaling process of IL-12A in innate immunity requires nitric oxide synthase 2A. Along with EBI3, IL-12A forms a novel heterodimeric cytokine, designated as interleukin-35 (IL-35), that may be specifically produced by T reg cells and is required for maximal suppressive activity.Specifications
IL-12 p35 | |
0.5 mg/mL | |
ELISA | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
P29459 | |
3592 | |
Primary | |
Affinity Chromatography |
B-T21 (BT21) | |
4°C | |
Monoclonal | |
Liquid | |
IgG1 | |
Human | |
IL12A | |
CLMF; CLMF p35; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; il 12; IL-12 subunit p35; IL-12, subunit p35; Il12a; IL-12A; IL12p35; IL-12p35; IL35 subunit; ILN; Interleukin; Interleukin 12; interleukin 12 p35 subunit; interleukin 12, p35; interleukin 12a; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); Interleukin12; interleukin-12 alpha chain; interleukin-12 beta p35 subunit; interleukin-12 p35 subunit; interleukin-12 subunit alpha; Ll12a; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NKSF1; P35 | |
IL12A | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.