Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-12 p35 Recombinant Superclonal™ Antibody (2H6L2)
IL-12 p35 Recombinant Polyclonal Antibody (2H6L2)
Supplier: Thermo Scientific 710154
Description
IL-12 p35 Recombinant Polyclonal Antibody for Western Blot
Specifications
| IL-12 p35 | |
| Recombinant Superclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide | |
| P29459, P48091 | |
| IL12A | |
| Recombinant protein corresponding to amino acids 23-328 of monkey IL-12. | |
| 100 μg | |
| Primary | |
| Monkey | |
| Antibody | |
| IgG |
| Western Blot | |
| 2H6L2 | |
| Unconjugated | |
| IL12A | |
| CLMF; CLMF p35; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; il 12; IL-12 subunit p35; IL-12, subunit p35; Il12a; IL-12A; IL12p35; IL-12p35; IL35 subunit; ILN; Interleukin; Interleukin 12; interleukin 12 p35 subunit; interleukin 12, p35; interleukin 12a; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); Interleukin12; interleukin-12 alpha chain; interleukin-12 beta p35 subunit; interleukin-12 p35 subunit; interleukin-12 subunit alpha; Ll12a; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NKSF1; P35 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3592, 703205 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction