Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12 p40 Mouse anti-Human, Unconjugated, Clone: 11B10, MilliporeSigma™

Catalog No. MABF22951MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF22951MI 100 μg
MABF22952MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF22951MI Supplier MilliporeSigma Supplier No. MABF2295100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-12 p40 Monoclonal antibody specifically detects IL-12 p40 in Human samples. It is validated for Immunohistochemistry (Paraffin), Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-12 p40
Applications Immunohistochemistry (Paraffin), Flow Cytometry
Classification Monoclonal
Clone 11B10
Conjugate Unconjugated
Gene Accession No. NP_002178.2
Gene Alias Interleukin-12 subunit beta;IL-12B;Cytotoxic lymphocyte maturation factor 40 kDa subunit;CLMF p40;IL-12 subunit p40;NK cell stimulatory factor chain 2;NKSF2
Gene Symbols IL12B, NKSF2
Host Species Mouse
Immunogen His-tagged full-length human recombinant Interleukin-12 subunit beta (IL12 p40).
Molecular Weight of Antigen 37.17 kDa calculated.
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 11B10 is a mouse monoclonal antibody that targets human Interleukin-12 p40 subunit.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.