Learn More
IL-12 p40 Mouse anti-Human, Unconjugated, Clone: 11B10, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | IL-12 p40 |
| Applications | Immunohistochemistry (Paraffin), Flow Cytometry |
| Classification | Monoclonal |
| Clone | 11B10 |
| Conjugate | Unconjugated |
| Gene Accession No. | NP_002178.2 |
| Gene Alias | Interleukin-12 subunit beta;IL-12B;Cytotoxic lymphocyte maturation factor 40 kDa subunit;CLMF p40;IL-12 subunit p40;NK cell stimulatory factor chain 2;NKSF2 |
| Gene Symbols | IL12B, NKSF2 |
| Host Species | Mouse |
| Immunogen | His-tagged full-length human recombinant Interleukin-12 subunit beta (IL12 p40). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.