Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12 (p40/p70) Rat anti-Mouse, PE, Clone: C15.6, BD

Catalog No. BDB562038 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562038 25 μg
BDB554479 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB562038 Supplier BD Biosciences Supplier No. 562038
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The C15.6 monoclonal antibody specifically binds to both free and complexed (homodimer p80 and heterodimer p70) forms of the p40 subunit of mouse interleukin-12 (IL-12). The immunogen used to generate the C15.6 hybridoma was recombinant mouse IL-12 p70 protein. p40 has also been described as a subunit of IL-23 and thus it is possible that the C15.6 antibody will crossreact with IL-23.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-12 (p40/p70)
Applications Flow Cytometry
Classification Monoclonal
Clone C15.6
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen CHO-expressed recombinant mouse IL-12 p70 protein
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.