Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-12 p40 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547197
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547197 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547197 Supplier Invitrogen™ Supplier No. PA547197
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, approximatley 50% cross-reactivity with recombinant mouse IL-12 is observed, 5% cross-reactivity with recombinant human IL-12 is observed, and less than 10% cross-reactivity with recombinant feline IL-12 and recombinant canine IL-12 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.30 EU per 1 μg of the antibody by the LAL method.

IL12B (IL-12 p40) encodes a subunit of interleukin 12, a cytokine that acts on T and natural killer cells, and has a broad array of biological activities. Interleukin 12 is a disulfide-linked heterodimer composed of the 40 kDa cytokine receptor like subunit encoded by this gene, and a 35 kDa subunit encoded by IL12A. This cytokine is expressed by activated macrophages that serve as an essential inducer of Th1 cells development. This cytokine has been found to be important for sustaining a sufficient number of memory/effector Th1 cells to mediate long-term protection to an intracellular pathogen. Overexpression of this gene was observed in the central nervous system of patients with multiple sclerosis (MS), suggesting a role of this cytokine in the pathogenesis of the disease. The promoter polymorphism of this gene has been reported to be associated with the severity of atopic and non-atopic asthma in children.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-12 p40
Applications Neutralization, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Il12b
Gene Accession No. 0
Gene Alias CLMF; CLMF p35; CLMF p40; CLMF2; cytotoxic lymphocyte maturation factor 40 kDa subunit; Il12; IL12 p40; IL-12 p40 subunit; IL-12 subunit p35; IL-12 subunit p40; IL12, subunit p40; IL-12.p40; IL12A; IL-12A; Il12b; IL-12B; Il12p40; IL-12p40; IL-12p40 subunit; IMD28; IMD29; Interleukin; interleukin 12 p40 subunit; interleukin 12 subunit p40; interleukin 12, p40; interleukin 12B; interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleukin-12 beta chain; interleukin-12 beta chain precursor; interleukin-12 p40; interleukin-12 p40 chain; interleukin-12 p40 subunit; interleukin-12 subunit beta; interleukin-12 subunit p40; interleukin-12p40; interleukin-23 p40 subunit; natural killer cell stimulatory factor, 40 kD subunit; NK cell stimulatory factor chain 2; NKSF; NKSF1; NKSF2; p40; RP23-388G23.1
Gene Symbols Il12b
Host Species Goat
Immunogen Sf 21-derived recombinant rat IL-12 p40: Met23-Ser335; p35: Arg23-Ser215.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64546
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.