Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-12 p70/IL-12A Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298685MFV450
Description
IL-12 p70/IL-12A Polyclonal antibody specifically detects IL-12 p70/IL-12A in Mouse samples. It is validated for Western BlotSpecifications
IL-12 p70/IL-12A | |
Polyclonal | |
Western Blot | |
CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35 | |
Produced in rabbits immunized with purified, a synthetic peptide corresponding to the N-terminus of the mouse IL-12 p70/IL-12A. | |
0.1 mL | |
Alzheimers Research, Immunology, Innate Immunity, Neurodegeneration | |
3592 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction