Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-12 p70/IL-12A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | IL-12 p70/IL-12A |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 3-5 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224627SS
![]() |
Novus Biologicals
NBP224627SS |
0.025 mg |
Each for $181.90
|
|
|||||
NBP224627
![]() |
Novus Biologicals
NBP224627 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
IL-12 p70/IL-12A Polyclonal specifically detects IL-12 p70/IL-12A in Human samples. It is validated for Western Blot.Specifications
IL-12 p70/IL-12A | |
Western Blot 3-5 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
3592 | |
A portion of amino acids 200-219 of IL-12A was used as the immunogen for this antibody. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Primate | |
CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35 | |
IL12A | |
IgG | |
Protein A purified | |
The amino acid sequence used as immunogen for this antibody is 100% homologous in human, monkey and gorilla and 95% homologous in mouse and dog. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title