Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-12 R beta 1 Antibody (69310), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB839Z
This item is not returnable.
View return policy
Description
IL-12 R beta 1 Monoclonal antibody specifically detects IL-12 R beta 1 in Human samples. It is validated for Flow CytometrySpecifications
IL-12 R beta 1 | |
Monoclonal | |
DyLight 755 | |
CD212, CD212 antigen, IL-12 receptor beta component, IL-12 receptor subunit beta-1, IL12R, IL-12R subunit beta-1, IL12RB, IL-12RB1, IL-12R-beta-1, IL-12R-BETA1, interleukin 12 receptor, beta 1, interleukin-12 receptor beta-1 chain, interleukin-12 receptor subunit beta-1, MGC34454 | |
Mouse myeloma cell line NS0-derived recombinant human IL-12 R beta 1, Cys24-Glu540, Accession # P42701 | |
0.5 mL | |
Cell Cycle and Replication | |
3594 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry | |
69310 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction