Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-12 R beta 2 Antibody (093), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290421APC
Description
IL-12 R beta 2 Monoclonal antibody specifically detects IL-12 R beta 2 in Mouse samples. It is validated for ELISASpecifications
IL-12 R beta 2 | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
093 | |
ELISA | |
IL-12 receptor beta 2, IL-12 receptor subunit beta-2, IL-12R subunit beta-2, IL-12RB2, IL-12R-beta-2, interleukin 12 receptor, beta 2, interleukin-12 receptor beta-2 chain, interleukin-12 receptor subunit beta-2 | |
Rabbit immunized with purified, recombinant Mouse IL-12 R beta 2 (Accession#: NP_032380.1, Met1-Asn637) | |
0.1 mL | |
Cell Cycle and Replication | |
3595 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction