Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-12B Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | IL-12B |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1702
![]()
|
Proteintech Group Inc
176451AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Interleukin-12 (IL-12) is a heterodimeric 70 kD (p70) cytokine composed of two covalently linked, glycosylated chains, 40kD (p40) and 35-kD (p35). IL-12 is mainly produced by monocytes, macrophages, and dendritic cells in response to bacterial products such as lipopolysaccharides (LPS), to intracellular pathogens or upon interaction with activated T cells. IL-12 was originally discovered because of its ability to induce interferon-gamma production, cell proliferation, and cytotoxicity mediated by natural killer cells and T cells. It is now established that IL-12 also plays a key role in the development of Th1 responses, leading to IFN-gamma and IL-2 production. These cytokines can in turn promote T-cell responses and macrophage activation. The p40 and p35 subunits by themselves have no IL-12 bioactivity, though the p40 homodimer has been shown to bind the IL-12 receptor and to be an antagonist of IL-12 p70. Free p40 is typically secreted in vast excess of IL-12 p70 by cells co-expressing both the p35 and p40 subunits. The p40 subunit of IL-12 has been shown to have extensive amino acid sequence homology to the extracellular domain of the IL-6 receptor, while the p35 subunit shows some homology to IL-6 and G-CSF.Specifications
| IL-12B | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P29460 | |
| 3593 | |
| IL-12B Fusion Protein Ag11837 | |
| Primary | |
| -20°C | |
| IL12B |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CLMF, CLMF p40, CLMF2, IL 12 subunit p40, IL 12B, IL12B, Interleukin 12 subunit beta, NKSF, NKSF2 | |
| IL12B | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title