Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-13 Antibody (13A), FITC, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP143239F
Description
IL-13 Monoclonal antibody specifically detects IL-13 in Mouse samples. It is validated for Flow Cytometry, ELISASpecifications
IL-13 | |
Monoclonal | |
FITC | |
PBS | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, ELISA | |
13A | |
Flow Cytometry, ELISA | |
ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600 | |
The immunogen for this antibody was IL13. | |
0.1 mL | |
Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
3596 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction