Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-13 Antibody (31606), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2131Y
Description
IL-13 Monoclonal antibody specifically detects IL-13 in Human samples. It is validated for Western Blot, Neutralization, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
IL-13 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Neutralization, Flow Cytometry (Intracellular Staining), CyTOF | |
31606 | |
Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600 | |
E. coli-derived recombinant human IL-13, Gly35-Asn146, Accession # P35225 | |
0.1 mL | |
Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
3596 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction