Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-13 Antibody (B-B13), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314573PE
Description
IL-13 Monoclonal antibody specifically detects IL-13 in Human samples. It is validated for Flow Cytometry, ELISA, Functional AssaySpecifications
IL-13 | |
Monoclonal | |
PE | |
ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600 | |
Recombinant human IL-13 | |
0.1 mL | |
Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
3596 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry, ELISA, Functional Assay | |
B-B13 | |
PBS | |
Mouse | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction