Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-13 Monoclonal Antibody (eBio1316H), Biotin, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5012181
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-121-81 50 μg
50-121-82 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-121-81 Supplier Invitrogen™ Supplier No. 13713581
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: The eBio1316H antibody reacts with mouse IL-13. The eBio1316H antibody is a neutralizing antibody. Mouse interleukin-13 (IL-13), originally designated p600, is a 11.6 kDa protein secreted by activated T helper cells, CD8+ T cells, and NK cells. IL-13 was originally identified in 1989 through differential hybridization of cDNA libraries prepared from T helper 1 and T helper 2 cells. IL-13 contains four cysteine residues, which form two intramolecular disulfide bonds. IL-13 shares many activities with IL-4; this relates to a requirement for IL-4Ralpha chain for signal transduction. The biological activities of IL-13 include suppression of macrophage cytotoxic activity, upregulation of IL-1RA expression, and suppression of inflammatory cytokine expression. IL-13 induces CD23 expression on B cells and enhances CD72, and expression of class II MHC gene products. Mouse IL-13 does not display the activities on T cells and B cell stimulatory effects which IL-4 exhibits. Mouse IL-13 is active on human and mouse cells. Applications Reported: The eBio1316H antibody has been reported useful for neutralization, ELISA detection, and ELISPOT detection. Applications Tested: The biotinylated eBio1316H antibody has been tested as the detection antibody in a sandwich ELISA (and ELISPOT assay) for analysis of mouse interleukin-13 (IL-13) in combination with the purified eBio13A (14-7133) antibody for capture and recombit mouse IL-13 (14-8131) as the standard.

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-13
Applications ELISA
Classification Monoclonal
Clone eBio1316H
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Il13
Gene Accession No. P20109
Gene Alias ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600
Gene Symbols Il13
Host Species Rat
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16163
Target Species Mouse
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.