Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-13 Monoclonal Antibody (eBio13A), Brilliant Ultra Violet™ 805, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 368713382
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-871-3382 100 μg
36-871-3380 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-871-3382 Supplier Invitrogen™ Supplier No. 368713382
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

The eBio13A antibody reacts with mouse IL-13. IL-13 is a cytokine produced mainly by Th2 cells, but also by antigen-primed CD8 T cells. IL-13 has a strong involvement in allergic inflammation and parasitic clearing and in cancer models has been shown to have either inhibitory or stimulatory activity depending on the tumor. In humans, IL-13 is found to play a role in isotype switching in B cells. IL-13 is implicating in down modulating macrophage activity, through the reduction of pro-inflammatory cytokines (IL-1, IL-6, IL-8, IL-10, IL-12) Applications Reported: This eBio13A antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This eBio13A antibody has been tested by intracellular staining followed by flow cytometric analysis of stimulated mouse splenocytes. This may be used at less than or equal to 1.0 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Brilliant Ultraviolet 805 is a tandem dye that emits at 797 nm and is intended for use on cytometers equipped with an ultraviolet (355 nm) laser.

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-13
Applications Flow Cytometry
Classification Monoclonal
Clone eBio13A
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 805
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Il13
Gene Accession No. P20109
Gene Alias ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600
Gene Symbols Il13
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16163
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.