Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-13 Mouse anti-Non-human Primate, Clone: IL13-I, Novus Biologicals™

Mouse Monoclonal Antibody
$313.07 - $1125.76
Specifications
| Antigen | IL-13 |
|---|---|
| Clone | IL13-I |
| Concentration | 0.5 mg/ml |
| Applications | ELISA |
| Classification | Monoclonal |
Description
IL-13 Monoclonal antibody specifically detects IL-13 in Non-human Primate samples. It is validated for ELISA, ELISPOTSpecifications
| IL-13 | |
| 0.5 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Non-human Primate | |
| ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600 | |
| Recombinant human IL-13 | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| IL13-I | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 0.2 um filtered PBS | |
| 3596 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title