Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-13R alpha 1 Antibody (419718) [DyLight 755], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1462Z
Description
IL-13R alpha 1 Monoclonal antibody specifically detects IL-13R alpha 1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
IL-13R alpha 1 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
419718 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
bB128O4.2.1 (interleukin 13 receptor, alpha 1), Cancer/testis antigen 19, CD213A1, CD213a1 antigen, CT19, IL13 receptor alpha-1 chain, IL-13 receptor subunit alpha-1, IL13R, IL-13R subunit alpha-1, IL-13Ra, IL13RA, IL-13RA1, IL-13R-alpha-1, interleukin 13 receptor, alpha 1, interleukin-13 receptor subunit alpha-1, NR4 | |
Mouse myeloma cell line NS0-derived recombinant human IL-13 R alpha 1, Ala27-Thr343, Accession # P78552 | |
0.1 mL | |
CD Markers, Immunology, Innate Immunity, Signal Transduction | |
3597 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction