Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-13R alpha 2 Antibody (007), Alexa Fluor™ 647

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290890AF647
Description
IL-13R alpha 2 Monoclonal specifically detects IL-13R alpha 2 in Rat samples. It is validated for ELISA.Specifications
IL-13R alpha 2 | |
Monoclonal | |
Alexa Fluor 647 | |
cancer/testis antigen 19, CD213a2, CD213a2 antigen, CT19, IL-13 receptor subunit alpha-2, IL13BP, IL13R, IL-13R, IL-13R subunit alpha-2, IL-13RA2, IL-13R-alpha-2, interleukin 13 binding protein, interleukin 13 receptor alpha 2 chain, interleukin 13 receptor, alpha 2, interleukin-13 receptor subunit alpha-2, Interleukin-13-binding protein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat IL-13R alpha 2 (Q8VHK6; Met1-Lys336). | |
0.1 mL | |
Cancer | |
3598 | |
Rat | |
Purified |
ELISA | |
7 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Mouse IL13RA2 No cross-reactivity in ELISA with: Human IL5Ralpha Human IL4R Rat IL9R Human IL13RA1 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction