Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-13R alpha 2 Antibody (MM0367-11S23), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211663MFV610
Description
IL-13R alpha 2 Monoclonal antibody specifically detects IL-13R alpha 2 in Human samples. It is validated for Flow CytometrySpecifications
IL-13R alpha 2 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
MM0367-11S23 | |
Flow Cytometry | |
cancer/testis antigen 19, CD213a2, CD213a2 antigen, CT19, IL-13 receptor subunit alpha-2, IL13BP, IL13R, IL-13R, IL-13R subunit alpha-2, IL-13RA2, IL-13R-alpha-2, interleukin 13 binding protein, interleukin 13 receptor alpha 2 chain, interleukin 13 receptor, alpha 2, interleukin-13 receptor subunit alpha-2, Interleukin-13-binding protein | |
Human recombinant IL-13R alpha 2 (extracellular domain) | |
0.1 mL | |
Cancer | |
3598 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction