Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-13RA2 (CD213a2) Mouse anti-Human, APC, Clone: 47 (also known as47; mAb47; 47 mAb), BD Phrmingen™
Mouse Monoclonal Antibody
$132.00 - $273.50
Specifications
Antigen | IL-13RA2 (CD213a2) |
---|---|
Clone | 47 (also known as47; mAb47; 47 mAb) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC |
Description
IL-13RA2 (CD213a2) Monoclonal antibody specifically detects IL-13RA2 (CD213a2) in Human samples. It is validated for Flow cytometrySpecifications
IL-13RA2 (CD213a2) | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
CD213A2; IL-13 receptor subunit alpha-2; IL-13R; IL-13Rα2; cancer/testis antigen 19 | |
Human rhIL13Ra2hFc | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
47 (also known as47; mAb47; 47 mAb) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
3598 | |
IgG1 κ | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title