Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-13RA2 (CD213a2) Mouse anti-Human, BV711, Clone: 47 (also known as47; mAb47; 47 mAb), BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 756124
Description
IL-13RA2 (CD213a2) Monoclonal antibody specifically detects IL-13RA2 (CD213a2) in Human samples. It is validated for Flow cytometry
Specifications
IL-13RA2 (CD213a2) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
3598 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG1 κ |
Flow Cytometry | |
47 (also known as47; mAb47; 47 mAb) | |
Brilliant Violet 711 | |
CD213A2; IL-13 receptor subunit alpha-2; IL-13R; IL-13Rα2; cancer/testis antigen 19 | |
Human rhIL13Ra2hFc | |
50 μg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction