Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-15 Antibody (1048222) [DyLight 488], Novus Biologicals™
SDP

Catalog No. FAB2472K Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2472K 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FAB2472K Supplier Novus Biologicals Supplier No. FAB2472K
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-15 Monoclonal antibody specifically detects IL-15 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry

Specifications

Antigen IL-15
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 1048222
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias IL-15MGC9721, interleukin 15, interleukin-15
Host Species Mouse
Immunogen E. coli-derived human IL-15 protein, Asn49-Ser162, Accession # P40933.1
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3600
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.