Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-15 Antibody (111), Alexa Fluor™ 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289449AF350
Description
IL-15 Monoclonal specifically detects IL-15 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
IL-15 | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
111 | |
ELISA, Sandwich ELISA Detection | |
IL-15MGC9721, interleukin 15, interleukin-15 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-15 (NP_000576.1; Asn18-Ser162). | |
0.1 mL | |
Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immune System Diseases, Immunology, Innate Immunity | |
3600 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction